5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

F0939-1G F0939-5G
TCF0939-1GEA 67.94 CAD
TCF0939-1G TCF0939-5G
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Formula: C8H9NO3
Melting Pt: 283°C
MDL Number: MFCD06202342
CAS Number: 253870-02-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR