2,4,6-Tribromopyridine ≥97.0% (by GC)

Supplier: TCI America
Warning

T3375-1G T3375-5G
76220-592EA 202.24 CAD
76220-592 76220-594
2,4,6-Tribromopyridine ≥97.0% (by GC)
2,4,6-Tribromopyridine
Formula: C₅H₂Br₃N
Boiling Pt: 284°C
MDL Number: MFCD00234002
CAS Number: 2408-70-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR