4-Tetrahydrothiopyranone-1,1-dioxide ≥98.0% (by GC)

Supplier: TCI America

T3320-1G T3320-200MG
TCT3320-1GEA 327.22 CAD
TCT3320-1G TCT3320-200MG
4-Tetrahydrothiopyranone-1,1-dioxide ≥98.0% (by GC)
4-Tetrahydrothiopyranone-1,1-dioxide
Formula: C₅H₈O₃S
MDL Number: MFCD00460698
CAS Number: 17396-35-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR