1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: 4-Oxopyrrolidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Ethyl ester, 1-tert-Butyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate

B4658-1G B4658-5G
TCB4658-1GEA 52.93 CAD
TCB4658-1G TCB4658-5G
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate ≥97.0% (by HPLC, total nitrogen)
1-tert-Butyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate
Formula: C₁₂H₁₉NO₅
MW: 257.2830 g/mol
Boiling Pt: 120°C
Storage Temperature: Refrigerator
MDL Number: MFCD06656201
CAS Number: 146256-98-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 56.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR