5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T2984-1G T2984-5G
TCT2984-1GEA 127.35 CAD
TCT2984-1G TCT2984-5G
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥97.0% (by titrimetric analysis)
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride
Formula: C₇H₁₀ClNOS
Storage Temperature: Freezer
MDL Number: MFCD11111130
CAS Number: 115473-15-9

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR