Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America

E1051-1G E1051-5G
TCE1051-1GEA 88.48 CAD
TCE1051-1G TCE1051-5G
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate ≥98.0% (by GC)
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate
Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00030352
CAS Number: 2199-59-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR