1H-Pyrazolo[3,4-b]pyridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P2285-1G P2285-200MG
TCP2285-1GEA 221.2 CAD
TCP2285-1G TCP2285-200MG
1H-Pyrazolo[3,4-b]pyridine ≥98.0% (by HPLC, titration analysis)
1H-Pyrazolo[3,4-b]pyridine
Formula: C₆H₅N₃
Boiling Pt: 120°C
MDL Number: MFCD05663981
CAS Number: 271-73-8

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR