Ethyl-3-(pyridin-2-ylamino)propanoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

E1214-25G E1214-5G
TCE1214-25GEA 120.08 CAD
TCE1214-25G TCE1214-5G
Ethyl-3-(pyridin-2-ylamino)propanoate ≥98.0% (by titrimetric analysis)
Ethyl-3-(pyridin-2-ylamino)propanoate
Formula: C₁₀H₁₄N₂O₂
Boiling Pt: 125°C
MDL Number: MFCD09833621
CAS Number: 103041-38-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR