2-(Trifluoromethyl)pyridin-4-ol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
H1528-1G
H1528-5G
TCH1528-1GEA
162.74
CAD
TCH1528-1G
TCH1528-5G
2-(Trifluoromethyl)pyridin-4-ol ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)pyridin-4-ol
Formula:
C₆H₄F₃NO |
MDL Number:
MFCD01571202 CAS Number: 170886-13-2 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...