4-Chloropyrimidin-5-amine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2937-1G A2937-200MG
TCA2937-1GEA 379.2 CAD
TCA2937-1G TCA2937-200MG
4-Chloropyrimidin-5-amine ≥97.0% (by GC, titration analysis)
4-Chloropyrimidin-5-amine
Formula: C₄H₄ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD06412561
CAS Number: 54660-78-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR