2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

C3089-1G C3089-5G
TCC3089-1GEA 96.38 CAD
TCC3089-1G TCC3089-5G
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₃N₂O₂
MDL Number: MFCD06255171
CAS Number: 72587-15-6

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR