2,4-Dihydroxy-5-methoxypyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Methoxy-2,4-pyrimidinediol

D4599-1G D4599-5G
TCD4599-1GEA 118.97 CAD
TCD4599-1G TCD4599-5G
2,4-Dihydroxy-5-methoxypyrimidine ≥98.0% (by GC, titration analysis)
2,4-Dihydroxy-5-methoxypyrimidine
Formula: C₅H₆N₂O₃
MW: 142.1100 g/mol
Melting Pt: 344°C
MDL Number: MFCD00187903
CAS Number: 6623-81-0
EINECS: 229-580-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR