5-Nitro-6-methyluracil ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
M2506-1G
M2506-5G
TCM2506-1GEA
26.86
CAD
TCM2506-1G
TCM2506-5G
5-Nitro-6-methyluracil ≥95.0% (by HPLC, titration analysis)
5-Nitro-6-methyluracil
Formula:
C5H5N3O4 |
MDL Number:
MFCD00047362 CAS Number: 16632-21-6 EINECS: 240-688-1 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...