(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2730-1G A2730-200MG
TCA2730-1GEA 517.43 CAD
TCA2730-1G TCA2730-200MG
(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98.0% (by GC, titration analysis)
(4-Amino-2-methyl-5-pyrimidinyl)methanol
Formula: C₆H₉N₃O
MDL Number: MFCD00849239
CAS Number: 73-67-6
Merck Index: 14,09561

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR