tert-Butyl (2-hydroxyethyl)(methyl)carbamate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5685-1G
B5685-5G
TCB5685-1GEA
37.92
CAD
TCB5685-1G
TCB5685-5G
tert-Butyl (2-hydroxyethyl)(methyl)carbamate ≥98.0% (by GC)
tert-Butyl (2-hydroxyethyl)(methyl)carbamate
Formula:
C₈H₁₇NO₃ Boiling Pt: 84°C |
MDL Number:
MFCD03425857 CAS Number: 57561-39-4 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...