5-Amino-3-bromo-2-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Amino-3-bromo-2-picoline

A2996-1G A2996-5G
TCA2996-1GEA 270.18 CAD
TCA2996-1G TCA2996-5G
5-Amino-3-bromo-2-methylpyridine ≥98.0% (by GC, titration analysis)
5-Amino-3-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.0372 g/mol
MDL Number: MFCD08060462
CAS Number: 186593-43-1

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR