6-Amino-5-bromoquinoxaline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2913-25G A2913-5G
TCA2913-25GEA 742.6 CAD
TCA2913-25G TCA2913-5G
6-Amino-5-bromoquinoxaline ≥98.0% (by GC, titration analysis)
6-Amino-5-bromoquinoxaline
Formula: C₈H₆BrN₃
MDL Number: MFCD00799024
CAS Number: 50358-63-9

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR