4-Amino-3-chloropyridine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2788-1G
A2788-5G
TCA2788-1GEA
89.27
CAD
TCA2788-1G
TCA2788-5G
4-Amino-3-chloropyridine ≥97.0% (by GC, titration analysis)
4-Amino-3-chloropyridine
Formula:
C₅H₅ClN₂ |
MDL Number:
MFCD03984473 CAS Number: 19798-77-7 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...