Methyl-2-amino-5-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M2796-1G
M2796-5G
TCM2796-1GEA
115.34
CAD
TCM2796-1G
TCM2796-5G
Methyl-2-amino-5-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-5-hydroxybenzoate
Formula:
C₈H₉NO₃ |
MDL Number:
MFCD05863263 CAS Number: 1882-72-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 158.0 to 162.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...