3,5-Dibromoisonicotinaldehyde ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,5-Dibromo-4-formylpyridine, 3,5-Dibromo-4-pyridinecarboxaldehyde, 3,5-Dibromoisonicotinaldehyde

D4674-1G D4674-5G
TCD4674-1GEA 55.3 CAD
TCD4674-1G TCD4674-5G
3,5-Dibromoisonicotinaldehyde ≥97.0% (by GC)
3,5-Dibromoisonicotinaldehyde
Formula: C6H3Br2NO
MW: 264.9000 g/mol
MDL Number: MFCD06410681
CAS Number: 70201-42-2

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR