5-Fluoro-2-methylindole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1047-1G
F1047-5G
TCF1047-1GEA
219.62
CAD
TCF1047-1G
TCF1047-5G
5-Fluoro-2-methylindole ≥98.0% (by GC)
5-Fluoro-2-methylindole
Formula:
C9H8FN Boiling Pt: 120°C |
MDL Number:
MFCD02093649 CAS Number: 399-72-4 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...