2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

C2999-1G C2999-5G
TCC2999-1GEA 72.68 CAD
TCC2999-1G TCC2999-5G
2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥98.0% (by GC)
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Formula: C₆H₂ClF₃IN
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR