5-Bromo-1H-indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B4790-1G
B4790-5G
TCB4790-1GEA
200.66
CAD
TCB4790-1G
TCB4790-5G
5-Bromo-1H-indole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Bromo-1H-indole-3-carboxylic acid
Formula:
C₉H₆BrNO₂ Boiling Pt: 240°C |
MDL Number:
MFCD05664007 CAS Number: 10406-06-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...