(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B5344-1G B5344-5G
TCB5344-1GEA 112.18 CAD
TCB5344-1G TCB5344-5G
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride ≥98.0% (by HPLC, titration analysis)
(2S,3aS,6aS)-Benzyl octahydrocyclopenta[b]pyrrole-2-carboxylate hydrochloride
Formula: C₁₅H₂₀ClNO₂
MDL Number: MFCD04974076
CAS Number: 87269-87-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -28.0 to -32.0 deg (C=1, DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR