5-Chloro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Chloro-7-azaindole

C2996-1G C2996-5G
TCC2996-1GEA 51.19 CAD
TCC2996-1G TCC2996-5G
5-Chloro-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD06659676
CAS Number: 866546-07-8

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR