Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥98.0% (by GC)

Supplier: TCI America

E1152-1G E1152-5G
TCE1152-1GEA 94.8 CAD
TCE1152-1G TCE1152-5G
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥98.0% (by GC)
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate
Formula: C₁₁H₁₁F₃O₂
Boiling Pt: 105°C
MDL Number: MFCD07779351
CAS Number: 721-63-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR