Methyl-3-hydroxypicolinate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2973-1G M2973-5G
TCM2973-1GEA 142.2 CAD
TCM2973-1G TCM2973-5G
Methyl-3-hydroxypicolinate ≥98.0% (by GC)
Methyl-3-hydroxypicolinate
Formula: C₇H₇NO₃
MDL Number: MFCD00661311
CAS Number: 62733-99-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 75.0 to 79.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR