4-Methyloxazole-5-carboxylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M2663-1G M2663-200MG
TCM2663-1GEA 564.06 CAD
TCM2663-1G TCM2663-200MG
4-Methyloxazole-5-carboxylic acid ≥98.0% (by titrimetric analysis)
4-Methyloxazole-5-carboxylic acid
Formula: C₅H₅NO₃
MDL Number: MFCD01571070
CAS Number: 2510-32-9

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR