5-Phenylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
P2347-1G
P2347-250MG
TCP2347-1GEA
237
CAD
TCP2347-1G
TCP2347-250MG
5-Phenylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Phenylthiophene-2-carboxylic acid
Formula:
C11H8O2S |
MDL Number:
MFCD01313432 CAS Number: 19163-24-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 188.0 to 192.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...