2-Chloro-5-fluoropyrimidin-4-one ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Chloro-5-fluoro-4-pyrimidinone

C3381-1G C3381-5G
TCC3381-1GEA 131.14 CAD
TCC3381-1G TCC3381-5G
2-Chloro-5-fluoropyrimidin-4-one ≥98.0% (by titrimetric analysis)
2-Chloro-5-fluoropyrimidin-4-one
Formula: C₄H₂ClFN₂O
MW: 148.5229 g/mol
MDL Number: MFCD01346689
CAS Number: 155-12-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR