2,4-Diamino-6-methoxypyrimidine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5088-1G D5088-5G
76219-852EA 237 CAD
76219-852 76219-854
2,4-Diamino-6-methoxypyrimidine ≥98.0% (by HPLC)
2,4-Diamino-6-methoxypyrimidine
Formula: C₅H₈N₄O
MW: 140.1400 g/mol
MDL Number: MFCD02091088
CAS Number: 3270-97-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR