Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
E1154-1G
E1154-5G
TCE1154-1GEA
148.84
CAD
TCE1154-1G
TCE1154-5G
Ethyl-2,4-dichloropyrimidine-5-carboxylate ≥98.0% (by GC)
Ethyl-2,4-dichloropyrimidine-5-carboxylate
Formula:
C₇H₆Cl₂N₂O₂ Boiling Pt: 145°C Storage Temperature: Refrigerator |
MDL Number:
MFCD09910281 CAS Number: 51940-64-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...