5-Bromo-2-methylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5266-1G B5266-5G
TCB5266-1GEA 67.94 CAD
TCB5266-1G TCB5266-5G
5-Bromo-2-methylpyrimidine ≥98.0% (by GC)
5-Bromo-2-methylpyrimidine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD07375143
CAS Number: 7752-78-5

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR