6-Quinolinecarboxaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Formylquinoline

Q0097-1G Q0097-5G
TCQ0097-1GEA 181.86 CAD
TCQ0097-1G TCQ0097-5G
6-Quinolinecarboxaldehyde ≥98.0% (by GC, titration analysis)
6-Quinolinecarboxaldehyde
Formula: C₁₀H₇NO
MW: 157.1700 g/mol
MDL Number: MFCD00805836
CAS Number: 4113-04-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR