2-Chloro-3-nitropyridin-4-amine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2798-1G
A2798-200MG
TCA2798-1GEA
311.26
CAD
TCA2798-1G
TCA2798-200MG
2-Chloro-3-nitropyridin-4-amine ≥97.0% (by GC, titration analysis)
2-Chloro-3-nitropyridin-4-amine
Formula:
C₅H₄ClN₃O₂ |
MDL Number:
MFCD07368898 CAS Number: 2789-25-5 |
Specification Test Results
Appearance | White to Yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...