1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2650-1G C2650-5G
TCC2650-1GEA 91.64 CAD
TCC2650-1G TCC2650-5G
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Formula: C₁₂H₈ClFN₂O₃
MDL Number: MFCD08458865
CAS Number: 100361-18-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 222.0 to 226.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR