tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

B4761-1G B4761-5G
TCB4761-1GEA 60.04 CAD
TCB4761-1G TCB4761-5G
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥98.0% (by GC)
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate
Formula: C₁₀H₁₄N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD01318456
CAS Number: 175463-32-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR