6-Bromoimidazo[1,2-a]pyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5599-1G B5599-5G
TCB5599-1GEA 88.48 CAD
TCB5599-1G TCB5599-5G
6-Bromoimidazo[1,2-a]pyrimidine ≥98.0% (by GC, titration analysis)
6-Bromoimidazo[1,2-a]pyrimidine
Formula: C₆H₄BrN₃
MDL Number: MFCD09261434
CAS Number: 865156-68-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR