7H-Pyrrolo[2,3-d]pyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 7-Deazapurine, 7H-Pyrrolo[2,3-d]pyrimidine

D4820-1G D4820-200MG
TCD4820-1GEA 216.46 CAD
TCD4820-1G TCD4820-200MG
7H-Pyrrolo[2,3-d]pyrimidine ≥98.0% (by GC, titration analysis)
7H-Pyrrolo[2,3-d]pyrimidine
Formula: C₆H₅N₃
MW: 119.1240 g/mol
MDL Number: MFCD08234403
CAS Number: 271-70-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR