Methyl-2-bromothiazole-5-carboxylate ≥95.0% (by GC)

Supplier: TCI America

M2669-1G M2669-5G
TCM2669-1GEA 74.26 CAD
TCM2669-1G TCM2669-5G
Methyl-2-bromothiazole-5-carboxylate ≥95.0% (by GC)
Methyl-2-bromothiazole-5-carboxylate
Formula: C₅H₄BrNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD03788565
CAS Number: 54045-74-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR