Ethyl-2-aminothiazole-4-carboxylate 98,0 GC, T
Supplier: TCI America
|
Warning
|
Synonyms:
Ethyl 2-aminothiazole-4-carboxylate
E1215-25G
E1215-5G
TCE1215-25GEA
194.34
CAD
TCE1215-25G
TCE1215-5G
Ethyl-2-aminothiazole-4-carboxylate 98,0 GC, T
Ethyl-2-aminothiazole-4-carboxylate
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol |
MDL Number:
MFCD00619079 CAS Number: 5398-36-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...