6-Phenylpicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2320-1G P2320-5G
TCP2320-1GEA 276.5 CAD
TCP2320-1G TCP2320-5G
6-Phenylpicolinic acid ≥98.0% (by GC, titration analysis)
6-Phenylpicolinic acid
Formula: C₁₂H₉NO₂
MDL Number: MFCD04114206
CAS Number: 39774-28-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR