Methyl-6-(hydroxymethyl)nicotinate ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2750-1G M2750-200MG
TCM2750-1GEA 336.54 CAD
TCM2750-1G TCM2750-200MG
Methyl-6-(hydroxymethyl)nicotinate ≥97.0% (by GC, titration analysis)
Methyl-6-(hydroxymethyl)nicotinate
Formula: C₈H₉NO₃
MDL Number: MFCD00643821
CAS Number: 56026-36-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR