5-Bromo-6-chloronicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4355-1G
B4355-5G
TCB4355-1GEA
56.88
CAD
TCB4355-1G
TCB4355-5G
5-Bromo-6-chloronicotinic acid ≥98.0% (by GC, titration analysis)
5-Bromo-6-chloronicotinic acid
Formula:
C₆H₃BrClNO₂ |
MDL Number:
MFCD01927098 CAS Number: 29241-62-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...