2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥98.0% (by GC)

Supplier: TCI America
Warning

H1602-1G H1602-5G
TCH1602-1GEA 96.38 CAD
TCH1602-1G TCH1602-5G
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol ≥98.0% (by GC)
2-Methyl-6-(trifluoromethyl)pyrimidin-4-ol
Formula: C₆H₅F₃N₂O
MDL Number: MFCD03265321
CAS Number: 2836-44-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR