Methyl-3-amino-6-bromopyrazine-2-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2655-1G M2655-5G
TCM2655-1GEA 77.42 CAD
TCM2655-1G TCM2655-5G
Methyl-3-amino-6-bromopyrazine-2-carboxylate ≥98.0% (by GC)
Methyl-3-amino-6-bromopyrazine-2-carboxylate
Formula: C₆H₆BrN₃O₂
MDL Number: MFCD00834964
CAS Number: 6966-01-4

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR