Methyl-2-bromothiazole-4-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M2615-1G M2615-5G
TCM2615-1GEA 77.42 CAD
TCM2615-1G TCM2615-5G
Methyl-2-bromothiazole-4-carboxylate ≥98.0% (by GC)
Methyl-2-bromothiazole-4-carboxylate
Formula: C₅H₄BrNO₂S
MDL Number: MFCD06659908
CAS Number: 170235-26-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR