a-Tosylbenzyl isocyanide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
T3157-1G
T3157-5G
TCT3157-1GEA
881.64
CAD
TCT3157-1G
TCT3157-5G
a-Tosylbenzyl isocyanide ≥98.0% (by HPLC, total nitrogen)
a-Tosylbenzyl isocyanide
Formula:
C₁₅H₁₃NO₂S |
MDL Number:
MFCD04114776 CAS Number: 36635-66-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...