1-(4-Chlorophenyl)-N-methylmethanamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 4-Chloro-N-methylbenzylamine

C2885-1G C2885-5G
TCC2885-1GEA 80.58 CAD
TCC2885-1G TCC2885-5G
1-(4-Chlorophenyl)-N-methylmethanamine ≥97.0% (by GC, titration analysis)
1-(4-Chlorophenyl)-N-methylmethanamine
Formula: C₈H₁₀ClN
MW: 155.6247 g/mol
Boiling Pt: 119°C
MDL Number: MFCD00018749
CAS Number: 104-11-0
EINECS: 203-175-3
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR