1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium hydrogensulfate ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI America
C3007-1G
C3007-5G
TCC3007-1GEA
129.56
CAD
TCC3007-1G
TCC3007-5G
1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium hydrogensulfate ≥95.0% (by HPLC, total nitrogen)
1-(((6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl)methyl)-6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium hydrogensulfate
Formula:
C₂₂H₂₄N₆O₉S₃ Melting Pt: 200°C Storage Temperature: Refrigerator |
MDL Number:
MFCD11864966 CAS Number: 98753-19-6 Merck Index: 14,01940 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (with Total Nitrogen) | min. 95.0 % |
Specific rotation [a]20/D | -27.0 to -33.0 deg (C=2.5,a solution prepared by addition of water to 25mL of actonitrile to make 50mL) (calcd.on anh.substance) |
Water | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...